missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Etoricoxib-d4, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
Etoricoxib D4 (MK-0663 D4) is a deuterium labeled Etoricoxib. Etoricoxib is a non steroidal anti-inflammatory agent, acting as a selective and orally active COX-2 inhibitor, with IC50s of 1.1 μM and 116 μM for COX-2 and COX-1 in human whole blood.
Specifications
Specifications
| Chemical Name or Material | Etoricoxib-d4 |
| CAS | 1131345-14-6 |
| Purity Grade Notes | Research |
| Physical Form | Powder |
| Molecular Formula | C18H11D4ClN2O2S |
| Color | Light Nude |
| Synonym | MK-0663-d4 L-791456-d4 |
| SMILES | ClC(C=N1)=CC(C2=C([2H])C([2H])=C(S(C)(=O)=O)C([2H])=C2[2H])=C1C3=CN=C(C)C=C3 |
| Molecular Weight (g/mol) | 362.87 |
| Quantity | 5 mg |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction