missing translation for 'onlineSavingsMsg'
Learn More

Etoposide, TRC

Product Code. 30012065
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012065 1 g 1g
18063140 50 mg 50mg
18072240 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012065 Supplier Toronto Research Chemicals Supplier No. TRCE9337501G
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33419-42-0
Molecular Formula C29 H32 O13
Molecular Weight (g/mol) 588.56
Synonym Etoposide, (5R,5aR,8aR,9S)-9-[[4,6-O-[(R)-Ethylidene]-β-d-glucopyranosyl]oxy]-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,8,8a,9-tetrahydroisobenzofuro[5,6-f][1,3]benzodioxol-6(5aH)-one, Furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one, 9-[[4,6-O-(1R)-ethylidene-β-D-glucopyranosyl]oxy]-5,8,8a,9-tetrahydro-5-(4-hydroxy-3,5-dimethoxyphenyl)-, (5R,5aR,8aR,9S)-, Epipodophyllotoxin, 4'-demethyl-, 4,6-O-ethylidene-β-D-glucopyranoside (8CI), Furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one, 9-[(4,6-O-ethylidene-β-D-glucopyranosyl)oxy]-5,8,8a,9-tetrahydro-5-(4-hydroxy-3,5-dimethoxyphenyl)-, [5R-[5α,5aβ,8aα,9β(R*)]]-, (5R,5aR,8aR,9S)-9-[[4,6-O-(1R)-Ethylidene-β-D-glucopyranosyl]oxy]-5,8,8a,9-tetrahydro-5-(4-hydroxy-3,5-dimethoxyphenyl)furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one, Pyrano[3,2-d]-1,3-dioxin, furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one deriv., (-)-Etoposide, 4'-Demethyl-1-O-[4,6-O-(ethylidene)-β-D-glucopyranosyl]epipodophyllotoxin, 4'-Demethylepipodophyllotoxin 9-(4,6-O-ethylidene-β-D-glucopyranoside), 4'-Demethylepipodophyllotoxin ethylidene-β-D-glucoside, Beposid, Bioposide, Celltop, EPE, Epipodophyllotoxin VP 16213, Eposed, Eposin, Ethopul, Eto-Gry, Etosid, Fytop, Fytosid, Lastet, NSC 141540, Nzytop, Sintopozid, Topok, Toposar, VP 16, VP 16 (pharmaceutical), VP 16-123, VP 16-213, VePesid, Vepesid J, Zuyeyidal, trans-Etoposide
IUPAC Name (5S,5aR,8aR,9R)-5-[[(2R,4aR,6R,7R,8R,8aS)-7,8-dihydroxy-2-methyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-6-yl]oxy]-9-(4-hydroxy-3,5-dimethoxyphenyl)-5a,6,8a,9-tetrahydro-5H-[2]benzofuro[5,6-f][1,3]benzodioxol-8-one
SMILES COc1cc(cc(OC)c1O)[C@H]2[C@@H]3[C@H](COC3=O)[C@H](O[C@@H]4O[C@@H]5CO[C@@H](C)O[C@H]5[C@H](O)[C@H]4O)c6cc7OCOc7cc26

Specifications

Quantity 1 g
InChI Formula InChI=1S/C29H32O13/c1-11-36-9-20-27(40-11)24(31)25(32)29(41-20)42-26-14-7-17-16(38-10-39-17)6-13(14)21(22-15(26)8-37-28(22)33)12-4-18(34-2)23(30)19(5-12)35-3/h4-7,11,15,20-22,24-27,29-32H,8-10H2,1-3H3/t11-,15+,20-,21-,22+,24-,25-,26-,27-,29+/m1/s1
Formula Weight 588.1843
Percent Purity >95%
Chemical Name or Material Etoposide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.