missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Etoposide Phosphate (Etopophos), Mikromol™
Description
Etoposide Phosphate (Etopophos)
Specifications
Specifications
| Chemical Name or Material | Etoposide Phosphate |
| CAS | 117091-64-2 |
| Analyte or Component Names | Etoposide Phosphate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C29 H33 O16 P |
| Certifications/Compliance | ISO 17034 |
| Synonym | Furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one, 5-[3,5-dimethoxy-4-(phosphonooxy)phenyl]-9-[[4,6-O-(1R)-ethylidene-β-D-glucopyranosyl]oxy]-5,8,8a,9-tetrahydro-, (5R,5aR,8aR,9S)-, Etoposide Phosphate, Etopofos, Etopophos |
| InChI Formula | InChI=1S/C29H33O16P/c1-11-38-9-20-27(42-11)23(30)24(31)29(43-20)44-25-14-7-17-16(40-10-41-17)6-13(14)21(22-15(25)8-39-28(22)32)12-4-18(36-2)26(19(5-12)37-3)45-46(33,34)35/h4-7,11,15,20-25,27,29-31H,8-10H2,1-3H3,(H2,33,34,35)/t11-,15+,20-,21-,22+,23-,24-,25-,27-,29+/m1/s1 |
| SMILES | COc1cc(cc(OC)c1OP(=O)(O)O)[C@H]2[C@@H]3[C@H](COC3=O)[C@H](O[C@@H]4O[C@@H]5CO[C@@H](C)O[C@H]5[C@H](O)[C@H]4O)c6cc7OCOc7cc26 |
| IUPAC Name | [4-[(5S,5aR,8aR,9R)-5-[[(2R,4aR,6R,7R,8R,8aS)-7,8-dihydroxy-2-methyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-6-yl]oxy]-8-oxo-5a,6,8a,9-tetrahydro-5H-[2]benzofuro[5,6-f][1,3]benzodioxol-9-yl]-2,6-dimethoxyphenyl] dihydrogen phosphate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?