missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Etoposide, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Etoposide
Specifications
Specifications
| Chemical Name or Material | Etoposide |
| CAS | 33419-42-0 |
| Analyte or Component Names | Etoposide |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C29 H32 O13 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Etoposide, Pyrano[3,2-d]-1,3-dioxin, furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one deriv., (-)-Etoposide, Beposid, Bioposide, Celltop, EPE, Epipodophyllotoxin VP 16213, Eposed, Eposin, Ethopul, Eto-Gry, Etosid, Fytop, Fytosid, Lastet, NSC 141540, Nzytop, Sintopozid, Topok, Toposar, VP 16, VP 16 (pharmaceutical), VP 16-123, VP 16-213, VePesid, Vepesid J, Zuyeyidal, trans-Etoposide, (5R,5aR,8aR,9S)-9-[[4,6-O-[(R)-Ethylidene]-beta-d-glucopyranosyl]oxy]-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,8,8a,9-tetrahydroisobenzofuro[5,6-f][1,3]benzodioxol-6(5aH)-one, Furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one, 9-[[4,6-O-(1R)-ethylidene-beta-D-glucopyranosyl]oxy]-5,8,8a,9-tetrahydro-5-(4-hydroxy-3,5-dimethoxyphenyl)-, (5R,5aR,8aR,9S)-, Epipodophyllotoxin, 4'-demethyl-, 4,6-O-ethylidene-beta-D-glucopyranoside (8CI), Furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one, 9-[(4,6-O-ethylidene-beta-D-glucopyranosyl)oxy]-5,8,8a,9-tetrahydro-5-(4-hydroxy-3,5-dimethoxyphenyl)-, [5R-[5alpha,5abeta,8aalpha,9beta(R*)]]-, (5R,5aR,8aR,9S)-9-[[4,6-O-(1R)-Ethylidene-beta-D-glucopyranosyl]oxy]-5,8,8a,9-tetrahydro-5-(4-hydroxy-3,5-dimethoxyphenyl)furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one, 4'-Demethyl-1-O-[4,6-O-(ethylidene)-beta-D-glucopyranosyl]epipodophyllotoxin, 4'-Demethylepipodophyllotoxin 9-(4,6-O-ethylidene-beta-D-glucopyranoside), 4'-Demethylepipodophyllotoxin ethylidene-beta-D-glucoside |
| InChI Formula | InChI=1S/C29H32O13/c1-11-36-9-20-27(40-11)24(31)25(32)29(41-20)42-26-14-7-17-16(38-10-39-17)6-13(14)21(22-15(26)8-37-28(22)33)12-4-18(34-2)23(30)19(5-12)35-3/h4-7,11,15,20-22,24-27,29-32H,8-10H2,1-3H3/t11-,15+,20-,21-,22+,24-,25-,26-,27-,29+/m1/s1 |
| SMILES | COc1cc(cc(OC)c1O)[C@H]2[C@@H]3[C@H](COC3=O)[C@H](O[C@@H]4O[C@@H]5CO[C@@H](C)O[C@H]5[C@H](O)[C@H]4O)c6cc7OCOc7cc26 |
| IUPAC Name | (5S,5aR,8aR,9R)-5-[[(2R,4aR,6R,7R,8R,8aS)-7,8-dihydroxy-2-methyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-6-yl]oxy]-9-(4-hydroxy-3,5-dimethoxyphenyl)-5a,6,8a,9-tetrahydro-5H-[2]benzofuro[5,6-f][1,3]benzodioxol-8-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction