missing translation for 'onlineSavingsMsg'
Learn More

Etonogestrel beta-D-Glucuronide, TRC

Product Code. 30381193
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.50mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381193 2.5 mg 2.50mg
30382042 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381193 Supplier Toronto Research Chemicals Supplier No. TRCE9336302.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Etonogestrel beta-D-Glucuronide
Analyte or Component Names Etonogestrel beta-D-Glucuronide
Shipping Condition Room Temperature
Molecular Formula C28 H36 O8
InChI Formula InChI=1S/C28H36O8/c1-4-27-13-14(3)20-17-9-7-16(29)12-15(17)6-8-18(20)19(27)10-11-28(27,5-2)36-26-23(32)21(30)22(31)24(35-26)25(33)34/h2,12,17-24,26,30-32H,3-4,6-11,13H2,1H3,(H,33,34)/t17-,18-,19-,20+,21-,22-,23+,24-,26-,27-,28-/m0/s1
Solution Type Neat
SMILES CC[C@]12CC(=C)[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@]2(O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)O)C#C
IUPAC Name (2S,3S,4S,5R,6S)-6-[[(8S,9S,10R,13S,14S,17R)-13-ethyl-17-ethynyl-11-methylidene-3-oxo-2,6,7,8,9,10,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 500.581
Quantity 2.5 mg
Formula Weight 500.241
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.