missing translation for 'onlineSavingsMsg'
Learn More

Etomoxir, MedChemExpress

Product Code. 16400538
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16400538

Brand: Medchem Express HY50202

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Etomoxir ((R)-(+)-Etomoxir) is an irreversible inhibitor of carnitine palmitoyltransferase 1a (CPT-1a), inhibits fatty acid oxidation (FAO) through CPT-1a and inhibits palmitate β-oxidation in human, rat and guinea pig.

Etomoxir ((R)-(+)-Etomoxir) is an irreversible inhibitor of carnitine palmitoyltransferase 1a (CPT-1a), inhibits fatty acid oxidation (FAO) through CPT-1a and inhibits palmitate β-oxidation in human, rat and guinea pig.

Specifications

Chemical Name or Material Etomoxir
CAS 124083-20-1
Color White
Purity Grade Notes Research
Molecular Formula C17H23ClO4
Quantity 5 mg
Synonym (R)-(+)-Etomoxir
Solubility Information DMSO : 140 mg/mL (428.37 mM; Need ultrasonic)
SMILES O=C(OCC)[C@@]1(OC1)CCCCCCOC2=CC=C(C=C2)Cl
Molecular Weight (g/mol) 326.82
Formula Weight 326.82
Percent Purity 99.4%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.