missing translation for 'onlineSavingsMsg'
Learn More

Etomidate hydrochloride, MedChemExpress

Product Code. 16478086
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16478086

Brand: Medchem Express HYB0100A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Etomidate hydrochloride (R 16659 hydrochloride) is a potent GABAA receptor agonist. Etomidate hydrochlorideis a neurological drug and effective parenteral medication and has the potential for management of endogenous hypercortisolaemia.

Etomidate hydrochloride (R 16659 hydrochloride) is a potent GABAA receptor agonist. Etomidate hydrochlorideis a neurological drug and effective parenteral medication and has the potential for management of endogenous hypercortisolaemia.

Specifications

Chemical Name or Material Etomidate hydrochloride
CAS 53188-20-8
Color White
Purity Grade Notes Research
Molecular Formula C14H17ClN2O2
Quantity 100 mg
Synonym R16659 hydrochloride
Solubility Information DMSO : ≥ 100 mg/mL (356.19 mM) ∣H2O : ≥ 50 mg/mL (178.09 mM)
SMILES O=C(C1=CN=CN1[C@@H](C2=CC=CC=C2)C)OCC.Cl
Molecular Weight (g/mol) 280.75
Formula Weight 280.75
Percent Purity 98.24%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.