missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Etomidate hydrochloride, MedChemExpress
Description
Etomidate hydrochloride (R 16659 hydrochloride) is a potent GABAA receptor agonist. Etomidate hydrochlorideis a neurological drug and effective parenteral medication and has the potential for management of endogenous hypercortisolaemia.
Specifications
Specifications
| Chemical Name or Material | Etomidate hydrochloride |
| CAS | 53188-20-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C14H17ClN2O2 |
| Quantity | 10 mg |
| Synonym | R16659 hydrochloride |
| Solubility Information | DMSO : ≥ 100 mg/mL (356.19 mM) ∣H2O : ≥ 50 mg/mL (178.09 mM) |
| SMILES | O=C(C1=CN=CN1[C@@H](C2=CC=CC=C2)C)OCC.Cl |
| Molecular Weight (g/mol) | 280.75 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?