missing translation for 'onlineSavingsMsg'
Learn More

Etomidate Acid, TRC

Product Code. 30383334
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383334 250 mg
30383067 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30383334 Supplier Toronto Research Chemicals Supplier No. TRCE933310250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56649-48-0
Molecular Formula C12 H12 N2 O2
Molecular Weight (g/mol) 216.24
Synonym 1-[(1R)-1-Phenylethyl]-1H-imidazole-5-carboxylic Acid, Etomidate Imp. A (EP) as R-Enantiomer, Etomidate Acid, 1H-Imidazole-5-carboxylic acid, 1-[(1R)-1-phenylethyl]-, 1H-Imidazole-5-carboxylic acid, 1-(1-phenylethyl)-, (R)-, (R)-(+)-1-(1-Phenylethyl)-1H-imidazole-5-carboxylic acid, 1-((1R)-1-Phenylethyl)-1H-imidazole-5-carboxylic acid, 1-(1R)-(1-Phenylethyl)-1H-imidazole-5-carboxylic acid, R 28141
IUPAC Name 3-[(1R)-1-phenylethyl]imidazole-4-carboxylic acid
SMILES C[C@H](c1ccccc1)n2cncc2C(=O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H12N2O2/c1-9(10-5-3-2-4-6-10)14-8-13-7-11(14)12(15)16/h2-9H,1H3,(H,15,16)/t9-/m1/s1
Formula Weight 216.0899
Percent Purity >95%
Chemical Name or Material Etomidate Acid (1-[(1R)-1-Phenylethyl]-1H-imidazole-5-carboxylic Acid)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.