missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Etomidate Acid (1-[(1R)-1-Phenylethyl]-1H-imidazole-5-carboxylic Acid), LoGiCal
Specifications
Specifications
| Chemical Name or Material | Etomidate Acid (1-[(1R)-1-Phenylethyl]-1H-imidazole-5-carboxylic Acid) |
| CAS | 56649-48-0 |
| Analyte or Component Names | Etomidate Acid (1-[(1R)-1-Phenylethyl]-1H-imidazole-5-carboxylic Acid) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H12 N2 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-[(1R)-1-Phenylethyl]-1H-imidazole-5-carboxylic Acid, Etomidate Imp. A (EP) as R-Enantiomer, Etomidate Acid, 1H-Imidazole-5-carboxylic acid, 1-[(1R)-1-phenylethyl]-, 1H-Imidazole-5-carboxylic acid, 1-(1-phenylethyl)-, (R)-, (R)-(+)-1-(1-Phenylethyl)-1H-imidazole-5-carboxylic acid, 1-((1R)-1-Phenylethyl)-1H-imidazole-5-carboxylic acid, 1-(1R)-(1-Phenylethyl)-1H-imidazole-5-carboxylic acid, R 28141, Etomidate Impurity A as R-Enantiomer |
| InChI Formula | InChI=1S/C12H12N2O2/c1-9(10-5-3-2-4-6-10)14-8-13-7-11(14)12(15)16/h2-9H,1H3,(H,15,16)/t9-/m1/s1 |
| Solution Type | Neat |
| SMILES | C[C@H](c1ccccc1)n2cncc2C(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?