missing translation for 'onlineSavingsMsg'
Learn More

Etofylline, TRC

Product Code. 30387526
Change view
Click to view available options
Quantity:
1 g
2.5 g
10 g
Unit Size:
2.50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387526 1 g -
30375075 2.5 g 2.50g
30369202 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387526 Supplier Toronto Research Chemicals Supplier No. TRCE9332001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 519-37-9
Molecular Formula C9 H12 N4 O3
Molecular Weight (g/mol) 224.22
Synonym 7-(2-Hydroxyethyl)-1,3-dimethyl-3,7-dihydro-1H-purine-2,6-dione, Etofylline, Diprophylline Imp. C (EP), Theophylline-Ethylenediamine Imp. F (EP), Theophylline Imp. F (EP), 3,7-Dihydro-7-(2-hydroxyethyl)-1,3-dimethyl-1H-purine-2,6-dione, 7-(β-Hydroxyethyl)theophylline, 7-(2-Hydroxyethyl)-1,3-dimethylxanthine, 7-(2-Hydroxyethyl)theophylline, 7-(Hydroxyethyl)theophylline, 7-Theophyllineethanol, Aethophyllinum, Ascorphylline, Bio-phylline, Cordalin, Corophyllin-N, Dilaphyllin, Etophyllin, Etophylline, Frekaphyllin, Hydroxyethyltheophylline, KT 200G, NSC 113373, Oxphylline, Oxyethophylline, Oxyethyltheophylline, Oxyphyllin, Oxyphylline, Oxytheonyl, Phyllocormin N, Soluphylline
IUPAC Name 7-(2-hydroxyethyl)-1,3-dimethylpurine-2,6-dione
SMILES CN1C(=O)N(C)c2ncn(CCO)c2C1=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H12N4O3/c1-11-7-6(8(15)12(2)9(11)16)13(3-4-14)5-10-7/h5,14H,3-4H2,1-2H3
Formula Weight 224.0909
Percent Purity >95%
Chemical Name or Material Etofylline
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.