missing translation for 'onlineSavingsMsg'
Learn More

Etofylline, Mikromol™

Product Code. 17981496
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17981496 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17981496 Supplier Mikromol Supplier No. MM0603.000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Etofylline
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Etofylline
CAS 519-37-9
Analyte or Component Names Etofylline
Shipping Condition Room Temperature
Molecular Formula C9 H12 N4 O3
Certifications/Compliance ISO 17034
Synonym Diprophylline Imp. C (EP), Theophylline Imp. F (EP), Theophylline-Ethylenediamine Anhydrous Imp. F (EP), Theophylline USP Related Compound F, Theophylline USP RC F, 7-(2-Hydroxyethyl)-1,3-dimethyl-3,7-dihydro-1H-purine-2,6-dione, Etofylline, Theophylline-Ethylenediamine Imp. F (EP), 3,7-Dihydro-7-(2-hydroxyethyl)-1,3-dimethyl-1H-purine-2,6-dione, 7-(beta-Hydroxyethyl)theophylline, 7-(2-Hydroxyethyl)-1,3-dimethylxanthine, 7-(2-Hydroxyethyl)theophylline, 7-(Hydroxyethyl)theophylline, 7-Theophyllineethanol, Aethophyllinum, Ascorphylline, Bio-phylline, Cordalin, Corophyllin-N, Dilaphyllin, Etophyllin, Etophylline, Frekaphyllin, Hydroxyethyltheophylline, KT 200G, NSC 113373, Oxphylline, Oxyethophylline, Oxyethyltheophylline, Oxyphyllin, Oxyphylline, Oxytheonyl, Phyllocormin N, Soluphylline, Theophylline Related Compound F, Diprophylline Impurity C, Theophylline Impurity F, Theophylline-Ethylenediamine Impurity F
InChI Formula InChI=1S/C9H12N4O3/c1-11-7-6(8(15)12(2)9(11)16)13(3-4-14)5-10-7/h5,14H,3-4H2,1-2H3
SMILES CN1C(=O)N(C)c2ncn(CCO)c2C1=O
IUPAC Name 7-(2-hydroxyethyl)-1,3-dimethylpurine-2,6-dione
Molecular Weight (g/mol) 224.22
Quantity 250 mg
Formula Weight 224.0909 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.