missing translation for 'onlineSavingsMsg'
Learn More

Etofibrate, MedChemExpress

Product Code. 16488298
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488298 10 mg 10mg
16463776 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488298 Supplier Medchem Express Supplier No. HYA0127

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Etofibrate is the ethandiol-1,2 diester of the nicotinic and clofibric acids. Etofibrate has been shown to be a potent hypolipidemic agent in animal and human.

Etofibrate is the ethandiol-1,2 diester of the nicotinic and clofibric acids. Etofibrate has been shown to be a potent hypolipidemic agent in animal and human.

Specifications

Chemical Name or Material Etofibrate
CAS 31637-97-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H18ClNO5
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (274.88 mM)
SMILES O=C(C1=CC=CN=C1)OCCOC(C(C)(OC2=CC=C(Cl)C=C2)C)=O
Molecular Weight (g/mol) 363.79
Formula Weight 363.79
Percent Purity 99.24%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.