missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ETNK1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110761
This item is not returnable.
View return policy
Description
Immunogen sequence: ANYIHVPPGS PEVPKLNVTV QDQEEHRCRE GALSLLQHLR PHWDPQEVTL QLFTDGITNK LIGCYVGNTM EDVVLVR Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 91%.
Ethanolamine kinase 1 (EKI1), functions in the first committed step of the phosphatidylethanolamine synthesis pathway. This cytosolic enzyme is specific for ethanolamine and exhibits negligible kinase activity on choline.
Specifications
| ETNK1 | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HBU6 | |
| ETNK1 | |
| Recombinant Protein corresonding to Human ETNK1. Recombinant protein control fragment (Product #RP-90290). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| ETNK1 | |
| 1110061E11Rik; 4930555L11Rik; AI195356; AI841245; C80956; D6Ertd3e; EKI; EKI 1; EKI1; Ethanolamine kinase; Ethanolamine kinase 1; Etnk1; I79_004753; Nbla10396; putative protein product of Nbla10396 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55500 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction