missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ETNK1 Monoclonal Antibody (3F11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA522149
This item is not returnable.
View return policy
Description
Peptide Sequence: WDPQEVTLQL FTDGITNKLI GCYVGNTMED VVLVRIYGNK TELLVDRDEE VKSFRVLQAH GCAPQLYCTF NNGLCYEFIQ GEALDPKHVC NPAIFR.
Ethanolamine kinase 1 (EKI1), functions in the first committed step of the phosphatidylethanolamine synthesis pathway. This cytosolic enzyme is specific for ethanolamine and exhibits negligible kinase activity on choline.
Specifications
| ETNK1 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9HBU6 | |
| ETNK1 | |
| ETNK1 (NP_061108, 133 a.a. ~ 228 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 3F11 | |
| Unconjugated | |
| ETNK1 | |
| 1110061E11Rik; 4930555L11Rik; AI195356; AI841245; C80956; D6Ertd3e; EKI; EKI 1; EKI1; Ethanolamine kinase; Ethanolamine kinase 1; Etnk1; I79_004753; Nbla10396; putative protein product of Nbla10396 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55500 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction