missing translation for 'onlineSavingsMsg'
Learn More

Etiracetam, MedChemExpress

Product Code. 16444237
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16444237 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16444237 Supplier Medchem Express Supplier No. HYB0106A
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Etiracetam (UCB 6474) is an acetylcholine agonist and a nootropic drug of the racetam family. Less active than its S-enantiomer Levetiracetam (UCB L059).

Etiracetam (UCB 6474) is an acetylcholine agonist and a nootropic drug of the racetam family. Less active than its S-enantiomer Levetiracetam (UCB L059).

Specifications

Chemical Name or Material Etiracetam
CAS 33996-58-6
Color White
Purity Grade Notes Research
Quantity 50 mg
Synonym UCB 6474
Solubility Information DMSO : ≥ 125 mg/mL (734.39 mM)
SMILES O=C(N)C(CC)N1C(CCC1)=O
Molecular Weight (g/mol) 170.21
Percent Purity 99.79%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.