missing translation for 'onlineSavingsMsg'
Learn More

Etimizol, MedChemExpress

Product Code. 16498547
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498547 5 mg 5mg
16486356 100 mg 100mg
16435918 10 mg 10mg
16422067 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498547 Supplier Medchem Express Supplier No. HY139185mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Etimizol(Ethymisole; Antiffine; Ethylnorantifein) was shown to relieve amnesia effectively in the origin of which there is the hypoxic component (hypobaric hypoxia, actinomycin D, mechanical injury of the brain).

Etimizol(Ethymisole

Specifications

Chemical Name or Material Etimizol
CAS 64-99-3
Color White
Purity Grade Notes Research
Molecular Formula C9H14N4O2
Quantity 5 mg
Synonym Ethimizole Ethymisol Ethymisole
Solubility Information Ethanol : ≥ 33.33 mg/mL (158.54 mM)
SMILES O=C(C1=C(C(NC)=O)N(CC)C=N1)NC
Molecular Weight (g/mol) 210.23
Formula Weight 210.23
Percent Purity 99.78%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.