missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Etilefrin Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Etilefrine hydrochloride |
| CAS | 943-17-9 |
| Molecular Formula | C10 H15 N O2 . Cl H |
| Quantity | 250 mg |
| Synonym | Benzenemethanol, α-[(ethylamino)methyl]-3-hydroxy-, hydrochloride (1:1), Benzenemethanol, α-[(ethylamino)methyl]-3-hydroxy-, hydrochloride (9CI), Benzyl alcohol, α-[(ethylamino)methyl]-m-hydroxy-, hydrochloride (6CI,8CI), (±)-Ethylphenylephrine hydrochloride, (±)-N-Ethylnorphenylephrine hydrochloride, 1-(3-Hydroxyphenyl)-2-ethylaminoethanol hydrochloride, Apocretin, Circupon, Effontil, Effortil, Effortilvet, Ethyl adrianol, Ethylephrine hydrochloride, Eti-Puren, Etilefrine hydrochloride, Funasol, Kertasin, Phetanol, Pulsamin, Tonus-Forte, Updormin, dl-1-(3-Hydroxyphenyl)-1-hydroxy-2-ethylaminoethane hydrochloride, dl-1-(3-Hydroxyphenyl)-2-ethylaminoethanol hydrochloride, dl-Effortil hydrochloride, dl-Ethylphenylephrine hydrochloride, dl-Etilefrin hydrochloride, dl-N-Ethylnorphenylephrine hydrochloride, α-[(Ethylamino)methyl]-m-hydroxybenzyl alcohol hydrochloride |
| InChI Formula | InChI=1S/C10H15NO2.ClH/c1-2-11-7-10(13)8-4-3-5-9(12)6-8;/h3-6,10-13H,2,7H2,1H3;1H |
| SMILES | Cl.CCNCC(O)c1cccc(O)c1 |
| IUPAC Name | 3-[2-(ethylamino)-1-hydroxyethyl]phenol;hydrochloride |
| Molecular Weight (g/mol) | 217.69 |
| Formula Weight | 217.087 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?