missing translation for 'onlineSavingsMsg'
Learn More

Etifoxine hydrochloride, MedChemExpress

Product Code. 16453127
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453127

Brand: Medchem Express HY16579

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Etifoxine hydrochloride, a non-benzodiazepine GABAergic compound, is a positive allosteric modulator of α1β2γ2 and α1β3γ2 subunit-containing GABAA receptors. Etifoxine hydrochloride reveals anxiolytic and anticonvulsant properties in rodents.

Etifoxine hydrochloride, a non-benzodiazepine GABAergic compound, is a positive allosteric modulator of α1β2γ2 and α1β3γ2 subunit-containing GABAA receptors. Etifoxine hydrochloride reveals anxiolytic and anticonvulsant properties in rodents.

Specifications

Chemical Name or Material Etifoxine hydrochloride
Physical Form Solid
CAS 56776-32-0
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C17H18Cl2N2O
For Use With (Application) Neuroscience-Neuromodulation
Synonym HOE 36-801 hydrochloride
Solubility Information DMSO : 50 mg/mL (148.26 mM; Need ultrasonic) ∣H2O : 5 mg/mL (14.83 mM; Need ultrasonic)
SMILES ClC1=CC=C2C(C(C3=CC=CC=C3)(C)OC(NCC)=N2)=C1.Cl
Molecular Weight (g/mol) 337.24
Formula Weight 337.24
Percent Purity 99.87%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.