missing translation for 'onlineSavingsMsg'
Learn More

Etidronic Acid-d3 Sodium Salt, TRC

Product Code. 30386626
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386626 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30386626 Supplier Toronto Research Chemicals Supplier No. TRCE9350025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Etidronic Acid-d3
CAS 774173-70-5
Molecular Formula C2 2H3 H5 O7 P2
Quantity 5 mg
Synonym Phosphonic acid, (1-hydroxyethylidene-2,2,2-d3)bis- (9CI), Etidronic Acid-d3, Etidronic Acid D3 (methyl D3)
InChI Formula InChI=1S/C2H8O7P2/c1-2(3,10(4,5)6)11(7,8)9/h3H,1H3,(H2,4,5,6)(H2,7,8,9)/i1D3
SMILES [2H]C([2H])([2H])C(O)(P(=O)(O)O)P(=O)(O)O
IUPAC Name (2,2,2-trideuterio-1-hydroxy-1-phosphonoethyl)phosphonic acid
Molecular Weight (g/mol) 209.05
Formula Weight 208.9934
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.