missing translation for 'onlineSavingsMsg'
Learn More

Etidronate disodium

Product Code. 15484769
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15484769 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15484769 Supplier Thermo Scientific Alfa Aesar Supplier No. J62381.03
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7414-83-7 | C2H6Na2O7P2 | 249.99 g/mol

Etidronate disodium is used as a bisphosphonate bone resorption inhibitor. Etidronic acid is used as a calcium regulator. Etidronic acid is a bisphosphonate antiresorptive agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Etidronate disodium is used as a bisphosphonate bone resorption inhibitor. Etidronic acid is used as a calcium regulator. Etidronic acid is a bisphosphonate antiresorptive agent.

Solubility
Soluble in water (26 mg/m
TRUSTED_SUSTAINABILITY

Specifications

CAS 7414-83-7
Melting Point >300°C
Color White
Molecular Formula C2H6Na2O7P2
MDL Number MFCD00152567
Quantity 1 g
Merck Index 14,3863
Synonym etidronate disodium, disodium etidronate, didronel, sodium etidronate, sodium ethidronate, sodium ethydronate, disodium ethydronate, didronel r, disodium ethanol-1,1-diphosphonate, etidronsaeure dinatriumsalz
Solubility Information Soluble in water (26mg/ml).
InChI Key GWBBVOVXJZATQQ-UHFFFAOYSA-L
SMILES [Na+].[Na+].CC(O)(P(O)([O-])=O)P(O)([O-])=O
IUPAC Name disodium hydrogen (1-hydrogen phosphonato-1-hydroxyethyl)phosphonate
Molecular Weight (g/mol) 249.99
PubChem CID 23894
ChEBI CHEBI:4906
Formula Weight 249.99
Physical Form Crystalline Solid
Chemical Name or Material Etidronate disodium
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.