missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Eticlopride hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Eticlopride hydrochloride is a selective dopamine D2/D3 receptor antagonist (Ki values are 0.50 and 0.16 nM respectively). Antipsychotic.
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-5-ethyl-N-[[(2S)-1-ethyl-2-pyrrolidinyl)methyl]-6-hydroxy-2-methoxy-benzamide hydrochloride |
| CAS | 97612-24-3 |
| Assay Percent Range | >99% |
| Quantity | 50 mg |
| Solubility | Soluble to 100mM in water |
| Molecular Formula | C17H25ClN2O3.HCl |
| Formula Weight | Observed MW: 377.31 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction