missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Eticlopride hydrochloride
GREENER_CHOICE

Product Code. 18033942
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18033942

Brand: Tocris Bioscience™ 1847/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D2 and D3 antagonist (D3 > D2)

Eticlopride hydrochloride is a selective dopamine D2/D3 receptor antagonist (Ki values are 0.50 and 0.16 nM respectively). Antipsychotic.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Chloro-5-ethyl-N-[[(2S)-1-ethyl-2-pyrrolidinyl)methyl]-6-hydroxy-2-methoxy-benzamide hydrochloride
CAS 97612-24-3
Assay Percent Range >99%
Quantity 50 mg
Solubility Soluble to 100mM in water
Molecular Formula C17H25ClN2O3.HCl
Formula Weight Observed MW: 377.31

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.