missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethynyltrimethylsilane, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Ethynyltrimethylsilane |
| CAS | 1066-54-2 |
| Molecular Formula | C5H10Si |
| Quantity | 25 g |
| Synonym | Trimethylsilylacetylene, (Trimethylsilanyl)acetylene, (Trimethylsilyl)acetylene, (Trimethylsilyl)ethyne, 1-(Trimethylsilyl)acetylene, 1-Trimethylsilylethyne, 2,2-Dimethyl-2-sila-3-butyne, Acetylenyltrimethylsilane, Ethynyltrimethylsilane, Trimethylethynylsilane, Trimethylsilanylethyne |
| InChI Formula | InChI=1S/C5H10Si/c1-5-6(2,3)4/h1H,2-4H3 |
| SMILES | C[Si](C)(C)C#C |
| IUPAC Name | ethynyl(trimethyl)silane |
| Molecular Weight (g/mol) | 98.22 |
| Formula Weight | 98.0552 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction