missing translation for 'onlineSavingsMsg'
Learn More

Ethynylferrocene (>90%), TRC

Product Code. 30386114
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386114 100 mg
30379642 500 mg
30387311 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386114 Supplier Toronto Research Chemicals Supplier No. TRCE937105100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1271-47-2
Molecular Formula C12H10Fe
Molecular Weight (g/mol) 210.06
Synonym Ferrocene, ethynyl- (8CI,9CI), Iron, cyclopentadienyl(ethynylcyclopentadienyl)- (7CI), 1-Ethynylferrocene, Ethynylferrocene, Ferrocenylacetylene, Ferrocenylethyne
IUPAC Name Iron, cyclopentadienyl(ethynylcyclopentadienyl)-
SMILES C#C[C-]12[CH]3=[CH]4[CH]5=[CH]1[Fe+2]67894532[CH]=%10[CH]9=[CH]8[CH-]7[CH]%106

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7H5.C5H5.Fe/c1-2-7-5-3-4-6-7;1-2-4-5-3-1;/h1,3-6H;1-5H;/q2*-1;+2
Formula Weight 210.01
Chemical Name or Material Ethynylferrocene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.