Learn More
Ethynylcyclopropane, 98%, Thermo Scientific Chemicals
£20.80 - £170.00
Chemical Identifiers
CAS | 6746-94-7 |
---|---|
Molecular Formula | C5H6 |
Molecular Weight (g/mol) | 66.1 |
MDL Number | MFCD02181090 |
InChI Key | NPTDXPDGUHAFKC-UHFFFAOYSA-N |
Synonym | cyclopropylacetylene, cyclopropyl acetylene, ethynyl cyclopropane, cyclopropane,ethynyl, cyclopropane, ethynyl, cyclopropylethyne, cyclopropyacetylene, ethynyl-cyclopropane, ethynylcyclo-propane, cyclopropylacethylene |
PubChem CID | 138823 |
IUPAC Name | ethynylcyclopropane |
SMILES | C#CC1CC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10744563
|
Thermo Scientific Acros
369130010 |
1 g | Glass bottle |
£20.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10354312
|
Thermo Scientific Acros
369130100 |
10 g | Glass bottle |
£52.50
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11954461
|
Thermo Scientific Acros
369130500 |
50 g | Glass bottle |
£170.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6746-94-7 | |
66.1 | |
NPTDXPDGUHAFKC-UHFFFAOYSA-N | |
138823 | |
C#CC1CC1 |
C5H6 | |
MFCD02181090 | |
cyclopropylacetylene, cyclopropyl acetylene, ethynyl cyclopropane, cyclopropane,ethynyl, cyclopropane, ethynyl, cyclopropylethyne, cyclopropyacetylene, ethynyl-cyclopropane, ethynylcyclo-propane, cyclopropylacethylene | |
ethynylcyclopropane |
Specifications
6746-94-7 | |
0.7801g/mL | |
−17°C | |
97.5% min. (GC) | |
1.4280 to 1.5300 | |
0.7801 | |
Solubility in water: 3.96g/L (20°C). Other solubilities: soluble in ethanol, dimethylsulfoxide, toluene,, acetone | |
C#CC1CC1 | |
66.1 | |
66.1 | |
Liquid |
Colorless to Yellow | |
51.0°C to 52.0°C | |
Authentic | |
C5H6 | |
MFCD02181090 | |
cyclopropylacetylene, cyclopropyl acetylene, ethynyl cyclopropane, cyclopropane,ethynyl, cyclopropane, ethynyl, cyclopropylethyne, cyclopropyacetylene, ethynyl-cyclopropane, ethynylcyclo-propane, cyclopropylacethylene | |
NPTDXPDGUHAFKC-UHFFFAOYSA-N | |
ethynylcyclopropane | |
138823 | |
98% | |
Ethynylcyclopropane, 98% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- Ethynylcyclopropane
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only