missing translation for 'onlineSavingsMsg'
Learn More

Ethynylboronic Acid MIDA Ester (>90%), TRC

Product Code. 30387048
Change view
Click to view available options
Quantity:
250 mg
2.5 g
5 g
Unit Size:
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387048 250 mg 250mg
30372020 5 g -
30373788 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387048 Supplier Toronto Research Chemicals Supplier No. TRCE932168250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1104637-53-7
Molecular Formula C7H8BNO4
Molecular Weight (g/mol) 180.95
Synonym Acetyleneboronic Acid MIDA Ester, Acetynylboronic Acid MIDA Ester, Ethyneboronic Acid MIDA Ester, (T-4)-[N-[(Carboxy-kO)methyl]-N-methylglycinato(2-)-kN,kO]ethynylboron
IUPAC Name 5-ethynyl-1-methyl-4,6-dioxa-1λ^{5}-aza-5λ^{4}-borabicyclo[3.3.0]octane-3,7-dione
SMILES C[N]12CC(=O)O[B]1(OC(=O)C2)C#C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C7H8BNO4/c1-3-8-9(2,4-6(10)12-8)5-7(11)13-8/h1H,4-5H2,2H3
Formula Weight 181.0546
Chemical Name or Material Ethynylboronic Acid MIDA Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.