missing translation for 'onlineSavingsMsg'
Learn More

Ethynyl Estradiol 3-Sulfate Sodium Salt (>85%), TRC

Product Code. 30383472
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383472 1 mg 1mg
30384803 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30383472 Supplier Toronto Research Chemicals Supplier No. TRCE6851151MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 724762-79-2
Molecular Formula C20H23NaO5S
Molecular Weight (g/mol) 398.45
Synonym (17α)-19-Norpregna-1,3,5(10)-trien-20-yne-3,17-diol 3-(Hydrogen Sulfate) Sodium Salt, 17α-Ethynylestradiol 3-Sulfate, Ethynylestradiol Sulfate Sodium Salt
SMILES [H][C@@]12CCC3=CC(OS(=O)(O[Na])=O)=CC=C3[C@@]1([H])CC[C@]4(C)[C@](O)(C#C)CC[C@@]24[H]

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C20H24O5S.Na/c1-3-20(21)11-9-18-17-6-4-13-12-14(25-26(22,23)24)5-7-15(13)16(17)8-10-19(18,20)2;/h1,5,7,12,16-18,21H,4,6,8-11H2,2H3,(H,22,23,24);/q;+1/p-1/t16-,17-,18+,19+,20+;/m1./s1
Percent Purity >95%
Chemical Name or Material Ethynyl Estradiol 3-Sulfate Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.