missing translation for 'onlineSavingsMsg'
Learn More

Ethynyl Estradiol-13C2, TRC

Product Code. 18081610
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18081610 25 mg 25mg
18022610 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18081610 Supplier Toronto Research Chemicals Supplier No. TRCE68510125MG
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2483735-63-1
Molecular Formula 13C2 C18 H24 O2
Molecular Weight (g/mol) 298.3887
Synonym 19-Norpregna-1,3,5(10)-trien-20-yne-3,17-diol-20,21-13C2, (17α)- (ACI), (17α)-19-Norpregna-1,3,5(10)-trien-20-yne-3,17-diol-20,21-13C2 (ACI), (17α)-19-Norpregna-1,3,5(10)-trien-20-yne-20,21-13C2-3,17-diol, Ethynyl-13C2 estradiol, Ethinylestradiol (ethinyl-13C2)
IUPAC Name (8R,9S,13S,14S,17S)-17-(1,2-^{13}C_{2})ethynyl-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-3,17-diol
SMILES C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CC[C@@]2(O)[13C]#[13CH]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C20H24O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,5,7,12,16-18,21-22H,4,6,8-11H2,2H3/t16-,17-,18+,19+,20+/m1/s1/i1+1,3+1
Formula Weight 298.1843
Chemical Name or Material Ethynyl Estradiol-13C2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.