missing translation for 'onlineSavingsMsg'
Learn More

Ethylvanillin, MedChemExpress

Product Code. 16478526
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478526 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16478526 Supplier Medchem Express Supplier No. HYB0940
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Ethylvanillin is a flavorant, about three times as potent as vanillin and is used in the production of chocolate.

Ethylvanillin is a flavorant, about three times as potent as vanillin and is used in the production of chocolate.

Specifications

Chemical Name or Material Ethylvanillin
CAS 121-32-4
Color White
Purity Grade Notes Research
Molecular Formula C9H10O3
Quantity 100 mg
Solubility Information Ethanol : ≥ 100 mg/mL (601.79 mM) ∣DMSO : ≥ 100 mg/mL (601.79 mM)
SMILES O=CC1=CC=C(O)C(OCC)=C1
Molecular Weight (g/mol) 166.17
Formula Weight 166.17
Percent Purity 99.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.