missing translation for 'onlineSavingsMsg'
Learn More

Ethylurea, 98%

Product Code. 15453257
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453257 25 g 25g
11446956 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15453257 Supplier Thermo Scientific Alfa Aesar Supplier No. B24040.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 625-52-5 | C3H8N2O | 88.11 g/mol

N-Ethyl urea, is a versatile building block, that can be used in organic synthesis of various pharmaceutical and biologically active compounds, as nitrogen and carbonyl source.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Ethyl urea, is a versatile building block, that can be used in organic synthesis of various pharmaceutical and biologically active compounds, as nitrogen and carbonyl source.

Solubility
Soluble in methanol (25 mg/ml), and water (100 mg/ml).

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-52-5
Molecular Formula C3H8N2O
Molecular Weight (g/mol) 88.11
MDL Number MFCD00007953
InChI Key RYECOJGRJDOGPP-UHFFFAOYSA-N
Synonym 1-ethylurea, n-ethylurea, urea, ethyl, urea, 1-ethyl, polyurethane, urea, n-ethyl, ethyl urea, pellethane, athylisoharnstoff
PubChem CID 12254
IUPAC Name ethylurea
SMILES CCNC(N)=O

Specifications

Melting Point 88°C to 94°C
Quantity 25 g
Beilstein 1740512
Solubility Information Soluble in methanol (25mg/ml),and water (100mg/ml).
Formula Weight 88.11
Percent Purity 98%
Chemical Name or Material Ethylurea
compliance-icons compliance-icons
Product Identifier
  • Ethylurea
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Germ cell mutagenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Ethylurea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.