missing translation for 'onlineSavingsMsg'
Learn More

Ethyltriphenylphosphonium iodide, 98+%

Product Code. 11498157
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498157 25 g 25g
11408167 100 g 100g
11418167 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498157 Supplier Thermo Scientific Alfa Aesar Supplier No. A17533.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4736-60-1 | C20H20IP | 418.26 g/mol

Ethyltriphenylphosphonium iodide is involved in synthesis of diarylmethine derivatives, phosphonium salts, and bismuth(III) polynuclear halide complexes, asymmetric hydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyltriphenylphosphonium iodide is involved in synthesis of diarylmethine derivatives, phosphonium salts, and bismuth(III) polynuclear halide complexes, asymmetric hydrogenation.

Solubility
Soluble in water.

Notes
Light Sensitive & Hygroscopic. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4736-60-1
Molecular Formula C20H20IP
Molecular Weight (g/mol) 418.26
MDL Number MFCD00040352
InChI Key SLAFUPJSGFVWPP-UHFFFAOYSA-M
Synonym ethyltriphenylphosphonium iodide, phosphonium, ethyltriphenyl-, iodide, ethyl triphenylphosphonium iodide, phenylphosphonium ethyl iodide, unii-xxz2v239jn, xxz2v239jn, ethyltriphenylphosphanium iodide, phosphonium, ethyltriphenyl-, iodide 1:1, ethyl triphenyl phosphanium iodide, ethyl triphenyl phosphonium iodide
PubChem CID 78474
IUPAC Name ethyl(triphenyl)phosphanium;iodide
SMILES [I-].CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 164°C to 169°C
Quantity 25 g
UN Number UN2811
Beilstein 3659323
Sensitivity Hygroscopic; Light sensitive
Solubility Information Soluble in water.
Formula Weight 418.26
Percent Purity ≥98%
Chemical Name or Material Ethyltriphenylphosphonium iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.