missing translation for 'onlineSavingsMsg'
Learn More

Ethyltrimethoxysilane, 97%

Product Code. 11489767
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489767 100 g 100g
11479767 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489767 Supplier Thermo Scientific Alfa Aesar Supplier No. B22752.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5314-55-6 | C5H14O3Si | 150.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5314-55-6
Molecular Formula C5H14O3Si
Molecular Weight (g/mol) 150.249
MDL Number MFCD00051541
InChI Key SBRXLTRZCJVAPH-UHFFFAOYSA-N
Synonym silane, ethyltrimethoxy, ethyl trimethoxy silane, trimethoxyethylsilane, silane,ethyltrimethoxy, acmc-209l4b, ethyltrimethoxysilane, dow corning™ product 1-6321
PubChem CID 79204
IUPAC Name ethyl(trimethoxy)silane
SMILES CC[Si](OC)(OC)OC

Specifications

Boiling Point 123°C to 124°C
Quantity 100 g
UN Number UN1993
Flash Point 27°C (81°F)
Formula Weight 150.25
Percent Purity 97%
Chemical Name or Material Ethyltrimethoxysilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.