missing translation for 'onlineSavingsMsg'
Learn More

Ethyltrimethoxysilane, 97%, AcroSeal™

Product Code. 10646840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10646840 100 mL AcroSeal™ Glass Bottle 100mL
10057949 1 L AcroSeal™ Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10646840 Supplier Thermo Scientific Acros Supplier No. 433081000
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5314-55-6 | C5H14O3Si | 150.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5314-55-6
Molecular Formula C5H14O3Si
Molecular Weight (g/mol) 150.25
InChI Key SBRXLTRZCJVAPH-UHFFFAOYSA-N
Synonym silane, ethyltrimethoxy, ethyl trimethoxy silane, trimethoxyethylsilane, silane,ethyltrimethoxy, acmc-209l4b, ethyltrimethoxysilane, dow corning™ product 1-6321
PubChem CID 79204
IUPAC Name ethyl(trimethoxy)silane
SMILES CC[Si](OC)(OC)OC

Specifications

Color Colorless to Yellow
Density 0.9400g/mL
Flash Point 27°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.3830 to 1.3850 (20°C, 589nm)
Quantity 100 mL
Specific Gravity 0.94
Solubility Information Solubility in water: decomposes
Formula Weight 150.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyltrimethoxysilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyltrimethoxysilane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.