missing translation for 'onlineSavingsMsg'
Learn More

Ethyltrichlorosilane, 99%, AcroSeal™, ACROS Organics™

Product Code. 11935841 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
100mL
Product Code. Quantity Packaging unitSize
11935841 1 L Glass Bottle 1 L
10506651 100 mL Glass Bottle 100mL
2 options
This item is not returnable. View return policy

Product Code. 11935841

Brand: Thermo Scientific Acros 313510010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 115-21-9
Molecular Formula C2H5Cl3Si
Molecular Weight (g/mol) 163.497
InChI Key ZOYFEXPFPVDYIS-UHFFFAOYSA-N
Synonym ethyltrichlorosilane, trichloro ethyl silane, trichloroethylsilicane, silane, trichloroethyl, ethyl silicon trichloride, trichloroethylsilicon, silicane, trichloroethyl, ethyl trichlorosilane, silane, ethyltrichloro, c2h5sicl3
PubChem CID 8260
IUPAC Name trichloro(ethyl)silane
SMILES CC[Si](Cl)(Cl)Cl

Specifications

Color Colorless to Yellow
Density 1.2380g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula H5C2SiCl3
Refractive Index 1.4240 to 1.4260 (20°C, 589nm)
Quantity 1 L
Formula Weight 163.51
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Ethyltrichlorosilane, 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.