missing translation for 'onlineSavingsMsg'
Learn More

Ethylparaben, MedChemExpress

Product Code. 16428526
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16428526 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16428526 Supplier Medchem Express Supplier No. HYB0934
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Ethylparaben is the ethyl ester of p-hydroxybenzoic acid, used as an antifungal preservative.

Ethylparaben is the ethyl ester of p-hydroxybenzoic acid, used as an antifungal preservative.

Specifications

Chemical Name or Material Ethylparaben
Physical Form Solid
CAS 120-47-8
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C9H10O3
For Use With (Application) COVID-19-immunoregulation
Synonym Ethyl parahydroxybenzoate Ethyl 4-hydroxybenzoate
Solubility Information DMSO : 100 mg/mL (601.79 mM; Need ultrasonic)
SMILES O=C(OCC)C1=CC=C(O)C=C1
Molecular Weight (g/mol) 166.17
Formula Weight 166.17
Percent Purity 98.23%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.