missing translation for 'onlineSavingsMsg'
Learn More

Ethylmethyldichlorosilane, 97%, Thermo Scientific™

Product Code. 11905921 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
This item is not returnable. View return policy

Product Code. 11905921

Brand: Thermo Scientific Acros 432890250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4525-44-4
Color Colorless
Density 1.0630g/mL
Boiling Point 100.0°C
Flash Point 2°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C3H8Cl2Si
Refractive Index 1.4180 to 1.4200 (20°C, 589nm)
MDL Number MFCD00013607
Quantity 25 mL
Specific Gravity 1.063
Synonym ethylmethyldichlorosilane, methylethyldichlorosilane, dichloro ethyl methylsilane, silane, dichloroethylmethyl, dichloro ethyl methyl silane, unii-47rob570jw, acmc-20akvy, dichloro-ethyl-methyl-silane, dichloro ethyl ?methylsilane, dichloro-ethyl-methyl silane
InChI Key PNECSTWRDNQOLT-UHFFFAOYSA-N
SMILES CC[Si](C)(Cl)Cl
IUPAC Name dichloro-ethyl-methylsilane
Molecular Weight (g/mol) 143.08
PubChem CID 78286
Formula Weight 143.09
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethylmethyldichlorosilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethylmethyldichlorosilane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P232-Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.