Learn More
Alfa Aesar™ Ethylmercury chloride
Brand: Alfa Aesar™ 037114.03
Code : NEW
Additional Details : CAS Number : 107-27-7 Weight : 0.00100kg Transport : UN number : 2025 Chem class : 1 Pack group : II
Quantity | 1g |
---|
Chemical Identifiers
Ethylmercury chloride | |
CH3CH2HgCl | |
MFCD00013593 | |
14,3826 |
107-27-7 | |
265.13 | |
UN2025 | |
Chloroethylmercury; Ethyl mercuric chloride |
Specifications
Ethylmercury chloride | |
1g | |
3.482g/mL | |
Crystalline | |
265.13 | |
UN2025 | |
Chloroethylmercury; Ethyl mercuric chloride |
193°C | |
107-27-7 | |
White | |
CH3CH2HgCl | |
MFCD00013593 | |
14,3826 | |
Insoluble in water; Slightly soluble in ether; Soluble in hot alcohol; Sublimes readily |