missing translation for 'onlineSavingsMsg'
Learn More

Ethylmercury Chloride (Technical Grade), TRC

Product Code. 30240860
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240860 25 mg 25mg
30240485 100 mg 100mg
30240636 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240860 Supplier Toronto Research Chemicals Supplier No. TRCE67955525MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-27-7
Molecular Formula C2 H5 Cl Hg
Molecular Weight (g/mol) 265.1
Synonym Ethylmercury chloride (6CI), Ceresan, Ceresan (fungicide), Chloroethylmercury, Criptodin, Cryptodin, Cryptodine, Emcon D, Ethylmercuric chloride, Granosan, Monoethylmercury chloride, NSC 1203, NSC 60515, Ethylmercury Chloride, Mercury, chloroethyl- (8CI,9CI)
IUPAC Name chloro(ethyl)mercury
SMILES CC[Hg]Cl

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C2H5.ClH.Hg/c1-2;;/h1H2,2H3;1H;/q;;+1/p-1
Formula Weight 265.9786
Chemical Name or Material Ethylmercury-chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.