missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylmalonic acid, 97+%
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Description
Ethylmalonic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethylmalonic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in water, methanol (very faint turbidity), and ethanol.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Ethylmalonic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in water, methanol (very faint turbidity), and ethanol.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 112°C to 114°C |
| Quantity | 25 g |
| Beilstein | 774334 |
| Solubility Information | Soluble in water,methanol (very faint turbidity),and ethanol. |
| Formula Weight | 132.12 |
| Percent Purity | 98% |
| Chemical Name or Material | Ethylmalonic acid |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction