missing translation for 'onlineSavingsMsg'
Learn More

Ethylmalonic acid, 97+%

Product Code. 11485566
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11485566

Brand: Thermo Scientific Alfa Aesar B20715.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 601-75-2 | C5H8O4 | 132.115 g/mol

Ethylmalonic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylmalonic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water, methanol (very faint turbidity), and ethanol.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 601-75-2
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.115
MDL Number MFCD00002668
InChI Key UKFXDFUAPNAMPJ-UHFFFAOYSA-N
Synonym ethylmalonic acid, 2-ethylmalonic acid, propanedioic acid, ethyl, 1,1-propanedicarboxylic acid, alpha-carboxybutyric acid, malonic acid, ethyl, ethylmalonate, ethyl-malonic acid, unii-432nf49dfg, ethylpropanedioic acid
PubChem CID 11756
ChEBI CHEBI:741548
IUPAC Name 2-ethylpropanedioic acid
SMILES CCC(C(=O)O)C(=O)O

Specifications

Melting Point 112°C to 114°C
Quantity 5 g
Beilstein 774334
Solubility Information Soluble in water,methanol (very faint turbidity),and ethanol.
Formula Weight 132.12
Percent Purity 98%
Chemical Name or Material Ethylmalonic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.