missing translation for 'onlineSavingsMsg'
Learn More

Ethylmagnesium bromide, 3M in diethyl ether, AcroSeal™

Product Code. p-130944 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10601425 100 mL AcroSeal™ Glass bottle 100mL
10703563 800 mL AcroSeal™ Glass bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10601425 Supplier Thermo Scientific Acros Supplier No. 347291000
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 925-90-6 | C2H5BrMg | 133.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 925-90-6 , 60-29-7
Molecular Formula C2H5BrMg
Molecular Weight (g/mol) 133.27
MDL Number MFCD00000043
InChI Key TWTWFMUQSOFTRN-UHFFFAOYSA-M
Synonym ethylmagnesium bromide, ethyl magnesium bromide, magnesium, bromoethyl, ethylmagnesiumbromide, bromoethylmagnesium, ethylmagnesium bromide solution, 1.0 m in thf, ethylmagnesium bromide solution, 3.0 m in diethyl ether, etmgbr, ethylmagnesiumbromid, ethylmagnesium bromid, grignard reagent
PubChem CID 101914
IUPAC Name magnesium;ethane;bromide
SMILES CC[Mg]Br

Specifications

Color Brown
Density 1.0300g/mL
Flash Point −40°C
Packaging AcroSeal™ Glass bottle
Linear Formula C2H5MgBr
Quantity 100 mL
Fieser 02,205
Specific Gravity 1.03
Formula Weight 133.27
Physical Form Liquid
Chemical Name or Material Ethylmagnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethylmagnesium bromide, 3M in Diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which, in contact with water, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P223-Do not allow contact with water.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.