missing translation for 'onlineSavingsMsg'
Learn More

Ethylidenecyclopentane, 90+%, Thermo Scientific Chemicals

Product Code. p-7024938
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11479656

Brand: Thermo Scientific Alfa Aesar B20209.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used to make other organic chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to make other organic chemicals.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2146-37-4
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00019319
InChI Key VONKRKBGTZDZNV-UHFFFAOYSA-N
Synonym cyclopentane, ethylidene, ethyl idenecyclopentane, ethylidene-cyclopentane, cyclopentane,ethylidene, acmc-1cs2b, 1e-ethylidenecyclopentane, ethylidenecyclopentane, 90+%
PubChem CID 75068
IUPAC Name ethylidenecyclopentane
SMILES CC=C1CCCC1

Specifications

Melting Point -130°C
Density 0.8
Boiling Point 112°C to 114°C
Refractive Index 1.45
Quantity 5 g
UN Number UN3295
Solubility Information Soluble in water.
Formula Weight 96.17
Percent Purity ≥90%
Chemical Name or Material Ethylidenecyclopentane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.