missing translation for 'onlineSavingsMsg'
Learn More

Ethylidene diacetate, 99%

Product Code. 10137280 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10137280 25 g Glass bottle 25g
10197370 100 g Glass bottle 100g
11914921 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10137280 Supplier Thermo Scientific Acros Supplier No. 410130250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 542-10-9 | C6H10O4 | 146.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-10-9
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.14
InChI Key ACKALUBLCWJVNB-UHFFFAOYSA-N
Synonym ethylidene diacetate, 1,1-diacetoxyethane, 1,1-ethanediol diacetate, 1,1-ethanediol, diacetate, ethylidene acetate, ethane-1,1-diyl diacetate, polyoxymethylenes, delrin, unii-kl1s8v6w25, 1-acetoxyethyl acetate
PubChem CID 222536
IUPAC Name 1-acetyloxyethyl acetate
SMILES CC(OC(=O)C)OC(=O)C

Specifications

Color Colorless to Yellow
Boiling Point 69.0°C (17.0 mmHg)
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OCOCH3)2
Refractive Index 1.3995 to 1.4015
Quantity 25 g
Merck Index 15,3866
Formula Weight 146.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethylidene diacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.