missing translation for 'onlineSavingsMsg'
Learn More

Ethylhexyl triazone, MedChemExpress

Product Code. 16481586
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481586 500 mg 500mg
16455236 100 mg 100mg
16401139 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16481586 Supplier Medchem Express Supplier No. HY109655
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ethylhexyl triazone is an approved ultraviolet-B (UV-B) chemical filter for commercial sunscreens.

Ethylhexyl triazone is an approved ultraviolet-B (UV-B) chemical filter for commercial sunscreens.

Specifications

Chemical Name or Material Ethylhexyl triazone
Physical Form Solid
CAS 88122-99-0
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C48H66N6O6
Synonym Octyl triazone
Solubility Information DMSO : 30 mg/mL (36.45 mM; Need ultrasonic and warming)
SMILES CCCCC(CC)COC(C(C=C1)=CC=C1NC2=NC(NC3=CC=C(C(OCC(CC)CCCC)=O)C=C3)=NC(NC4=CC=C(C(OCC(CC)CCCC)=O)C=C4)=N2)=O
Molecular Weight (g/mol) 823.07
Formula Weight 823.07
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.