missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trans-beta-methylcinnamate, 97%, Thermo Scientific Chemicals

Catalog Number 10113254
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1504-72-9
C12H14O2
190.24
MFCD00053762
BSXHSWOMMFBMLL-MDZDMXLPSA-N
e-ethyl 3-phenylbut-2-enoate, ethyl 3-phenylbut-2-enoate, ethyl 3-phenylcrotonate, ethyl 2e-3-phenylbut-2-enoate, ethyl trans-beta-methylcinnamate, ethyl e-3-phenylbut-2-enoate, ethyl e-3-phenyl-2-butenoate, ethyl trans-, a-methylcinnamate, 2-butenoic acid, 3-phenyl-, ethyl ester, cinnamic acid, .beta.-methyl-, ethyl ester
5354836
ethyl (E)-3-phenylbut-2-enoate
CCOC(=O)C=C(C)C1=CC=CC=C1
This item is not returnable. View return policy
1504-72-9
C12H14O2
190.24
MFCD00053762
BSXHSWOMMFBMLL-MDZDMXLPSA-N
e-ethyl 3-phenylbut-2-enoate, ethyl 3-phenylbut-2-enoate, ethyl 3-phenylcrotonate, ethyl 2e-3-phenylbut-2-enoate, ethyl trans-beta-methylcinnamate, ethyl e-3-phenylbut-2-enoate, ethyl e-3-phenyl-2-butenoate, ethyl trans-, a-methylcinnamate, 2-butenoic acid, 3-phenyl-, ethyl ester, cinnamic acid, .beta.-methyl-, ethyl ester
5354836
ethyl (E)-3-phenylbut-2-enoate
CCOC(=O)C=C(C)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.0420g/mL
Boiling Point 142°C to 144°C (20.0 hPa)
Flash Point >113°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C(CH3)=CHCO2C2H5
Quantity 1 g
Specific Gravity 1.042
Formula Weight 190.24
Percent Purity 97%
Chemical Name or Material Ethyl trans-β-methylcinnamate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'