missing translation for 'onlineSavingsMsg'
Learn More

Ethyl thiooxamate, 95%, Thermo Scientific Chemicals

Product Code. 10237363 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10237363 5 g Glass bottle 5g
10472651 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10237363 Supplier Thermo Scientific Acros Supplier No. 319600050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16982-21-1
Molecular Formula C4H7NO2S
Molecular Weight (g/mol) 133.17
MDL Number MFCD00074903
InChI Key YMBMCMOZIGSBOA-UHFFFAOYSA-N
Synonym ethyl 2-amino-2-thioxoacetate, ethyl carbamothioylformate, ethyl thioxamate, ethyl thiooxamidate, ethyl amino thioxo acetate, ethyl thiooxamat, ethyl aminothioxoacetate, ethyl 2-thiooxamate, acetic acid, aminothioxo-, ethyl ester, thiooxamic acid ethyl ester
PubChem CID 2733398
IUPAC Name ethyl 2-amino-2-sulfanylideneacetate
SMILES CCOC(=O)C(=S)N

Specifications

Melting Point 60.0°C to 66.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula H2NCSCO2C2H5
Quantity 5 g
Formula Weight 133.17
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl thiooxamate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.