missing translation for 'onlineSavingsMsg'
Learn More

ethylethiomorpholine-3-carboxylate hydrochloride, 90%, Thermo Scientific™

Product Code. p-3881337
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10255604 1 g Amber Glass Bottle 1g
11306835 5 g Amber Glass Bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10255604

Brand: Thermo Scientific Maybridge CC27673DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 159381-07-4
Molecular Formula C7H14ClNO2S
Molecular Weight (g/mol) 211.704
MDL Number MFCD00107968
InChI Key QQRCEZBURAQLBQ-UHFFFAOYSA-N
Synonym ethyl thiomorpholine-3-carboxylate hydrochloride, thiomorpholine-3-carboxylic acid ethyl ester hydrochloride, thiomorpholine-3-carboxylic acid ethyl ester hcl, thiomorpholine-3-carboxylicacidethylesterhydrochloride, ethyl thiomorpholine-3-carboxylate hcl, ethylthiomorpholine-3-carboxylatehydrochloride, 3-ethoxycarbonyl thiomorpholine hydrochloride, ethyl 1,4-thiazinane-3-carboxylate hydrochloride, ethyl 1,4-thiazaperhydroine-3-carboxylate, chloride, thiomorpholine-3-carboxylic acid ethyl esterhydrochloride
PubChem CID 17433447
IUPAC Name ethyl thiomorpholine-3-carboxylate;hydrochloride
SMILES CCOC(=O)C1CSCCN1.Cl

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 211.71
Percent Purity 90%
Chemical Name or Material ethylethiomorpholine-3-carboxylate hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.