missing translation for 'onlineSavingsMsg'
Learn More

Ethyl propionate, 99+%, Thermo Scientific Chemicals

Catalog Number 10183050
Click to view available options
Quantity:
25 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
105-37-3
C5H10O2
102.13
MFCD00009308
FKRCODPIKNYEAC-UHFFFAOYSA-N
ethyl propionate, propanoic acid, ethyl ester, propionic ester, propionic ether, propionic acid, ethyl ester, ethylpropionate, propionate d'ethyle, propanoic acid ethyl ester, ethyl n-propionate, propionic acid ethyl ester
7749
CHEBI:41330
ethyl propanoate
CCC(=O)OCC
This item is not returnable. View return policy
105-37-3
C5H10O2
102.13
MFCD00009308
FKRCODPIKNYEAC-UHFFFAOYSA-N
ethyl propionate, propanoic acid, ethyl ester, propionic ester, propionic ether, propionic acid, ethyl ester, ethylpropionate, propionate d'ethyle, propanoic acid ethyl ester, ethyl n-propionate, propionic acid ethyl ester
7749
CHEBI:41330
ethyl propanoate
CCC(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Acidity 1mg KOH/g max.
Melting Point -73°C
Color Colorless to Yellow
Density 0.8870g/mL
Boiling Point 99°C
Flash Point 12°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C2H5CO2C2H5
Refractive Index 1.3830 to 1.387
Quantity 500 mL
Beilstein 02, 240
Fieser 01,385
Merck Index 15, 3899
Specific Gravity 0.887
Solubility Information Solubility in water: 25g/L water (15°C). Other solubilities: miscible with alcohol,ether and propylene glycol
Viscosity <1 mPa.s (20°C)
Formula Weight 102.13
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Ethyl propionate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'