missing translation for 'onlineSavingsMsg'
Learn More

Ethyl oxamate, 99%

Product Code. 10209853 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
Packaging:
Glass bottle
Unit Size:
25g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10209853 500 g Glass bottle 500g
10549600 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10209853 Supplier Thermo Scientific Acros Supplier No. 118795000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 617-36-7 | C4H7NO3 | 117.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-36-7
MDL Number MFCD00008005
InChI Key RZMZBHSKPLVQCP-UHFFFAOYSA-N
Synonym ethyl oxamate, ethyl carbamoylformate, oxamic acid ethyl ester, ethyl amino oxo acetate, oxamethane, acetic acid, aminooxo-, ethyl ester, ethoxalamide, oxamidic acid ethyl ester, ethoxy carbonyl formamide, oxalic acid, monoethyl ester amide
PubChem CID 69238
IUPAC Name ethyl 2-amino-2-oxoacetate
SMILES CCOC(=O)C(=O)N

Specifications

Melting Point 112°C to 115°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min
Packaging Glass bottle
Molecular Formula C4H7NO3
Linear Formula H2NCOCO2C2H5
Quantity 500 g
Beilstein 02, 544
Solubility Information Solubility in water: soluble
Molecular Weight (g/mol) 117.1
Formula Weight 117.1
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Ethyl oxamate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.